Learn More
3-Bromo-4-chlorobenzotrifluoride, 97%, Thermo Scientific™
Product Code. 11430078
Change view
Click to view available options
Quantity:
5 g
10 g
50 g
Unit Size:
10g
50g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11430078 50 g 50g
11309519 5 g 5g
11420078 10 g 10g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Product Code. 11430078 Supplier Thermo Scientific Alfa Aesar Supplier No. B23666.18
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
missing translation for 'validMainframeMsg'
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 454-78-4
Molecular Formula C7H3BrClF3
Molecular Weight (g/mol) 259.45
MDL Number MFCD00018093
InChI Key APSISOSWYXCEQX-UHFFFAOYSA-N
Synonym 2-bromo-1-chloro-4-trifluoromethyl benzene, 3-bromo-4-chlorobenzotrifluoride, benzene, 2-bromo-1-chloro-4-trifluoromethyl, 3-bromo-4-chloro-alpha,alpha,alpha-trifluorotoluene, 4-chloro-3-bromobenzotrifluoride, 3-bromo-4-chloro-a,a,a-trifluorotoluene, pubchem4837, ksc915a8j, apsisoswyxceqx-uhfffaoysa, 3-bromo-4-chloro-benzotrifluoride
PubChem CID 67988
IUPAC Name 2-bromo-1-chloro-4-(trifluoromethyl)benzene
SMILES C1=CC(=C(C=C1C(F)(F)F)Br)Cl
Specifications
Melting Point -23°C to -22°C
Density 1.726
Boiling Point 191°C to 192°C
Flash Point 94°C (201°F)
Refractive Index 1.499
Quantity 50 g
Beilstein 1638470
Formula Weight 259.44
Percent Purity 97%
Chemical Name or Material 3-Bromo-4-chlorobenzotrifluoride