missing translation for 'onlineSavingsMsg'
Learn More

3-(Chloromethyl)-5-methylisoxazole, 97%, Thermo Scientific™

Product Code. 10525954
Change view
Click to view available options
Quantity:
250 mg
1 g
5 g
Unit Size:
1g
250mg
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
10525954 5 g 5g
10213192 250 mg 250mg
10631313 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 10525954 Supplier Thermo Scientific Maybridge Supplier No. SPB01262DE
 Restricted Item
Estimated Shipment: 23-10-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Chemical Identifiers

CAS 35166-37-1
Molecular Formula C5H6ClNO
Molecular Weight (g/mol) 131.56
MDL Number MFCD00221166
InChI Key FEXTXBAFBURKGS-UHFFFAOYSA-N
Synonym 3-chloromethyl-5-methylisoxazole, 3-chloromethyl-5-methyl-1,2-oxazole, 3-chloromethyl-5-methyl-isoxazole, pubchem8710, zlchem 1024, acmc-209idp, 3-chloromethyl-5-methyisoxazole, 3-choromethyl-5-methylisoxazole, 5-methyl-3-chloromethylisoxazole
PubChem CID 2735763
IUPAC Name 3-(chloromethyl)-5-methyl-1,2-oxazole
SMILES CC1=CC(CCl)=NO1

Specifications

Assay Percent Range 97%
Packaging Amber glass bottle
Quantity 5 g
Formula Weight 131.56
Percent Purity 97%
Chemical Name or Material 3-(Chloromethyl)-5-methylisoxazole
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.