Learn More
3-(Dimethylamino)-2-methyl-2-propenal, 99%, Thermo Scientific™
Product Code. 11915331 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
5 g
Unit Size:
5g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11915331 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 11915331 Supplier Thermo Scientific Acros Supplier No. 440370050
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
missing translation for 'validMainframeMsg'
Specifications
CAS 19125-76-9
CAS Min % 98.5
CAS Max % 100.0
Melting Point 34°C to 38°C
Color Yellow
Density 0.9710g/mL
Boiling Point 72°C to 74°C (0.3 mmHg)
Flash Point 113°C
Infrared Spectrum Authentic
Assay Percent Range 98.5% min. (GC)
Molecular Formula C6H11NO
Quantity 5 g
Specific Gravity 0.971
Synonym z-3-dimethylamino-2-methylacrylaldehyde, 2z-3-dimethylamino-2-methylacrylaldehyde, 2z-3-dimethylamino-2-methylprop-2-enal, z-3-dimethylamino-2-methylprop-2-enal, 2-propenal, 3-dimethylamino-2-methyl-, z, 2-propenal, 3-dimethylamino-2-methyl-, 2z
InChI Key JGAMOQYFDMQPRJ-XQRVVYSFSA-N
SMILES CC(=CN(C)C)C=O
IUPAC Name (Z)-3-(dimethylamino)-2-methylprop-2-enal
Molecular Weight (g/mol) 113.16
PubChem CID 5919306
Formula Weight 113.16
Percent Purity 99%
Physical Form Oil or Low Melting Solid
Chemical Name or Material 3-(Dimethylamino)-2-methyl-2-propenal
Show More Show Less
compliance-icons
Product Identifier
  • 3-(Dimethylamino)-2-methyl-2-propenal
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H315-Causes skin irritation.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.