missing translation for 'onlineSavingsMsg'
Learn More

3'-Fluoro-4'-hydroxyacetophenone, 98+%, Thermo Scientific™

Product Code. 11359915
Change view
Click to view available options
Quantity:
250 mg
1 g
Unit Size:
1g
250mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11359915 250 mg 250mg
11349915 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11359915 Supplier Thermo Scientific Alfa Aesar Supplier No. L19892.MD

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 403-14-5
Molecular Formula C8H7FO2
Molecular Weight (g/mol) 154.14
MDL Number MFCD00215840
InChI Key GSWTXZXGONEVJC-UHFFFAOYSA-N
Synonym 3'-fluoro-4'-hydroxyacetophenone, 1-3-fluoro-4-hydroxyphenyl ethanone, 1-3-fluoro-4-hydroxyphenyl-1-ethanone, 1-3-fluoro-4-hydroxyphenyl ethan-1-one, 3-fluoro-4-hydroxyacetophenone, ethanone, 1-3-fluoro-4-hydroxyphenyl, 1-3-fluoro-4-hydroxy-phenyl-ethanone, 1-acetyl-3-fluoro-4-hydroxybenzene, pubchem3492, acmc-20a0dy
PubChem CID 2737325
IUPAC Name 1-(3-fluoro-4-hydroxyphenyl)ethanone
SMILES CC(=O)C1=CC(=C(C=C1)O)F

Specifications

Melting Point 127°C to 129°C
Quantity 250 mg
Beilstein 1936529
Formula Weight 154.14
Percent Purity ≥98%
Chemical Name or Material 3'-Fluoro-4'-hydroxyacetophenone
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.