missing translation for 'onlineSavingsMsg'
Learn More

3-Formylthiophene-2-boronic acid, 97%

Product Code. 11318206
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11318206 5 g 5g
11308206 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11318206 Supplier Thermo Scientific Alfa Aesar Supplier No. L15196.06
 Restricted Item
Estimated Shipment: 23-07-2026
Add to basket
This item is not returnable. View return policy

CAS: 17303-83-2 | C5H5BO3S | 155.962 g/mol

3-Formylthiophene-2-boronic acid is used as reactant for Suzuki-Miyaura cross-coupling reactions.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
3-Formylthiophene-2-boronic acid is used as reactant for Suzuki-Miyaura cross-coupling reactions.

Solubility
Slightly soluble in water.

Notes
Air Sensitive, store away form air. Store in cool. Keep container tightly closed in a dry and well-ventilated place. Store away from oxidizing agent.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 17303-83-2
Molecular Formula C5H5BO3S
Molecular Weight (g/mol) 155.962
MDL Number MFCD01075679
InChI Key HYXMHAHVUFTVFZ-UHFFFAOYSA-N
Synonym 3-formylthiophen-2-yl boronic acid, 3-formylthiophene-2-boronic acid, 3-formyl-2-thienyl boronic acid, 3-formyl-2-thiopheneboronic acid, 3-formylthiophen-2-yl-2-boronic acid, 3-formyl-2-thienylboronic acid, 3-thiophenecarbaldehyde boronic acid, boronic acid, 3-formyl-2-thienyl, 3-formylthiophen-2-boronic acid, boronic acid,b-3-formyl-2-thienyl
PubChem CID 2773429
IUPAC Name (3-formylthiophen-2-yl)boronic acid
SMILES B(C1=C(C=CS1)C=O)(O)O

Specifications

Melting Point ∽180°C (decomposition)
Quantity 5 g
Beilstein 4387740
Sensitivity Air sensitive
Solubility Information Slightly soluble in water.
Formula Weight 155.97
Percent Purity 97%
Chemical Name or Material 3-Formylthiophene-2-boronic acid

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.