missing translation for 'onlineSavingsMsg'
Learn More

3'-Hydroxyacetophenone, 98%

Product Code. 11484234
Change view
Click to view available options
Quantity:
100 g
500 g
Unit Size:
100g
500g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11484234 100 g 100g
11494234 500 g 500g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11484234 Supplier Thermo Scientific Alfa Aesar Supplier No. A12547.22
 Restricted Item
In Stock
Add to basket
This item is not returnable. View return policy

CAS: 121-71-1 | C8H8O2 | 136.15 g/mol

3’Hydroxyacetophenone is used in the preparation of chalcones and flavonoids, which finds application as an anti-tuberculosis agent and antileishmanial chaocones. It is also involved in the preparation of phenylephrine. Further, it is an active component of castoreum, which is used as a tincture in some perfumes and as a food additive.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
3’Hydroxyacetophenone is used in the preparation of chalcones and flavonoids, which finds application as an anti-tuberculosis agent and antileishmanial chaocones. It is also involved in the preparation of phenylephrine. Further, it is an active component of castoreum, which is used as a tincture in some perfumes and as a food additive.

Solubility
Soluble in alcohol. Insoluble in water.

Notes
Incompatible with strong oxidizing agents and bases.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 121-71-1
Molecular Formula C8H8O2
Molecular Weight (g/mol) 136.15
MDL Number MFCD00002298
InChI Key LUJMEECXHPYQOF-UHFFFAOYSA-N
Synonym 3'-hydroxyacetophenone, 3-hydroxyacetophenone, 1-3-hydroxyphenyl ethanone, m-hydroxyacetophenone, 3-acetylphenol, 1-3-hydroxyphenyl ethan-1-one, ethanone, 1-3-hydroxyphenyl, m-acetylphenol, 3-hydroxy acetophenone, 3-acetophenol
PubChem CID 8487
IUPAC Name 1-(3-hydroxyphenyl)ethanone
SMILES CC(=O)C1=CC(=CC=C1)O

Specifications

Melting Point 93°C to 97°C
Density 1.1
Boiling Point 296°C
Quantity 100 g
Beilstein 2040676
Solubility Information Soluble in alcohol. Insoluble in water.
Formula Weight 136.15
Percent Purity 98%
Chemical Name or Material 3'-Hydroxyacetophenone
compliance-icons
Product Identifier
  • 3'-Hydroxyacetophenone
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/soap.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P312-Call a POISON CENTER/doctor/if you feel unwell.
Supplemental information
  • MIXTURE LIST-Contain: 1-(3-hydroxyphenyl)ethanone

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.