missing translation for 'onlineSavingsMsg'
Learn More

3-Methyl-2-benzothiazolinone Hydrazone Hydrochloride Hydrate, TRC

Product Code. 30469267
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
Unit Size:
1g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30469267 1 g 1g
30473271 25 g 25g
30478185 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30469267 Supplier Toronto Research Chemicals Supplier No. TRCM2889301G
 Restricted Item

This item requires a price update prior to being ordered.

Request a Price for this item here.

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 149022-15-1
Molecular Formula C8H10ClN3S.xH2O
Molecular Weight (g/mol) 233.718
Synonym 3-Methyl-2(3H)-benzothiazolone Hydrazone Monohydrochloride Hydrate, MBTH Hydrochloride Hydrate
IUPAC Name (Z)-2-hydrazineylidene-3-methyl-2,3-dihydrobenzo[d]thiazole hydrochloride hydrate
SMILES CN1/C(SC2=C1C=CC=C2)=N/N.O.Cl

Specifications

Quantity 1 g
InChI Formula InChI=1S/C8H9N3S.ClH.H2O/c1-11-6-4-2-3-5-7(6)12-8(11)10-9;;/h2-5H,9H2,1H3;1H;1H2/b10-8-;;
Formula Weight 233.039
Chemical Name or Material 3-Methyl-2-benzothiazolinone Hydrazone Hydrochloride Hydrate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.