Learn More
3-Nitro-o-xylene, 99%
Product Code. 11431407
Change view
Click to view available options
Quantity:
100 g
500 g
Unit Size:
100g
500g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11431407 100 g 100g
11441407 500 g 500g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 11431407 Supplier Thermo Scientific Alfa Aesar Supplier No. B24917.22
 Restricted Item
In Stock
Add to basket
CAS: 83-41-0 | C8H9NO2 | 151.17 g/mol
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 83-41-0
Molecular Formula C8H9NO2
Molecular Weight (g/mol) 151.17
MDL Number MFCD00007162
InChI Key FVHAWXWFPBPFOS-UHFFFAOYSA-N
Synonym 3-nitro-o-xylene, 2,3-dimethylnitrobenzene, nitroxylene, o-xylene, 3-nitro, benzene, 1,2-dimethyl-3-nitro, xylene, ar-nitro, benzene, dimethylnitro, unii-49gnx4cy5w, ccris 3117, 49gnx4cy5w
PubChem CID 6739
IUPAC Name 1,2-dimethyl-3-nitrobenzene
SMILES CC1=CC=CC(=C1C)[N+]([O-])=O
Specifications
Melting Point 13°C to 15°C
Density 1.129
Quantity 100 g
Refractive Index 1.544
UN Number UN1665
Beilstein 2045105
Boiling Point 244°C to 246°C
Formula Weight 151.17
Percent Purity 99%
Chemical Name or Material 3-Nitro-o-xylene