missing translation for 'onlineSavingsMsg'
Learn More

3-Phenoxybenzaldehyde, TRC

Product Code. 30005060
Change view
Click to view available options
Quantity:
50 mg
2.5 g
25 g
Unit Size:
2.5g
25g
50mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30005060 2.5 g 2.5g
30010842 50 mg 50mg
30007012 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30005060 Supplier Toronto Research Chemicals Supplier No. TRCP2978002.5G
 Restricted Item

This item requires a price update prior to being ordered.

Request a Price for this item here.

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 39515-51-0
Molecular Formula C13 H10 O2
Molecular Weight (g/mol) 198.22
Synonym Benzaldehyde, 3-phenoxy-, 3-Phenoxybenzaldehyde, 5-Phenoxybenzaldehyde, m-(Phenyloxy)benzaldehyde, m-Phenoxybenzaldehyde, m-Phenoxybenzaldehyde
IUPAC Name 3-phenoxybenzaldehyde
SMILES O=Cc1cccc(Oc2ccccc2)c1

Specifications

Quantity 2.5 g
InChI Formula InChI=1S/C13H10O2/c14-10-11-5-4-8-13(9-11)15-12-6-2-1-3-7-12/h1-10H
Formula Weight 198.0681
Percent Purity >95%
Chemical Name or Material 3-Phenoxybenzaldehyde
Grade HPLC
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.