Learn More
3-Phenylpyridine, 95%
Product Code. 11436796
Change view
Click to view available options
Quantity:
5 g
25 g
Unit Size:
25g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11436796 5 g 5g
11446796 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 11436796 Supplier Thermo Scientific Alfa Aesar Supplier No. B23930.06
Request Bulk or Custom Format
CAS: 1008-88-4 | C11H9N | 155.2 g/mol
3-Phenylpyridine is a useful synthetic intermediate. It is an important raw material and intermediate used in organic synthesis, pharmaceuticals, agrochemicals and dyestuff.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
3-Phenylpyridine is a useful synthetic intermediate. It is an important raw material and intermediate used in organic synthesis, pharmaceuticals, agrochemicals and dyestuff.
Solubility
Soluble in chloroform, dichloromethane and ethyl acetate.
Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Incompatible material is strong oxidizing agents.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 1008-88-4
Molecular Formula C11H9N
Molecular Weight (g/mol) 155.2
MDL Number MFCD00006380
InChI Key HJKGBRPNSJADMB-UHFFFAOYSA-N
Synonym pyridine, 3-phenyl, 3-phenyl-pyridine, m-phenylpyridine, unii-edz6kb7jvc, 5-phenylpyridine, 3-phenyipyridine, edz6kb7jvc, 3-azabiphenyl, 3-pyridylbenzene, 3-phenyl pyridine
PubChem CID 13886
IUPAC Name 3-phenylpyridine
SMILES C1=CC=C(C=C1)C2=CN=CC=C2
Specifications
Density 1.085
Boiling Point 272°C
Flash Point >110°C (230°F)
Refractive Index 1.616
Quantity 5 g
Beilstein 110400
Solubility Information Soluble in chloroform,dichloromethane and ethyl acetate.
Formula Weight 155.2
Percent Purity 97%
Chemical Name or Material 3-Phenylpyridine