Learn More
3-(Pyrid-2-yloxy)benzoic acid, 97%, Thermo Scientific™
Product Code. 10604975
Change view
Click to view available options
Quantity:
250 mg
1 g
Packaging:
Amber Glass Bottle
Unit Size:
1g
250mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
10604975 1 g Amber Glass Bottle 1g
10788075 250 mg Amber Glass Bottle 250mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 10604975 Supplier Thermo Scientific Maybridge Supplier No. CC50201DA
 Restricted Item
Estimated Shipment: 23-10-2026
Add to basket
missing translation for 'validMainframeMsg'
Chemical Identifiers
CAS 51362-30-2
Molecular Formula C12H9NO3
Molecular Weight (g/mol) 215.208
MDL Number MFCD09025853
InChI Key LYSIEAIIZBZRCE-UHFFFAOYSA-N
Synonym 3-pyridin-2-yloxy benzoic acid, 3-pyrid-2-yloxy benzoic acid, 3-2-pyridyloxy benzoic acid, 3-2 pyridyloxy-benzoic acid, 3-pyridin-2-yl oxy benzoic acid, benzoic acid, 3-2-pyridinyloxy
PubChem CID 21901423
IUPAC Name 3-pyridin-2-yloxybenzoic acid
SMILES C1=CC=NC(=C1)OC2=CC=CC(=C2)C(=O)O
Specifications
CAS Min % 94.5
CAS Max % 100.0
Assay Percent Range 95%
Packaging Amber Glass Bottle
Quantity 1 g
Formula Weight 215.21
Percent Purity 95%
Chemical Name or Material 3-(Pyrid-2-yloxy)benzoic acid