missing translation for 'onlineSavingsMsg'
Learn More

3-Pyridyl isothiocyanate, ≥95%, Thermo Scientific™

Product Code. 10324092
Change view
Click to view available options
Quantity:
1 g
10 g
Packaging:
Amber Glass Bottle
Unit Size:
10g
1g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10324092 1 g Amber Glass Bottle 1g
10653783 10 g Amber Glass Bottle 10g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10324092 Supplier Thermo Scientific Maybridge Supplier No. CC04107DA

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Chemical Identifiers

CAS 17452-27-6
Molecular Formula C6H4N2S
Molecular Weight (g/mol) 136.172
MDL Number MFCD00052351
InChI Key VMSZFBSYWXMXRF-UHFFFAOYSA-N
Synonym 3-pyridyl isothiocyanate, pyridine, 3-isothiocyanato, 3-isothiocyanato-pyridine, pyridine-3-isothiocyanate, pyridine,3-isothiocyanato, 3-pyridylisothiocyanate, 3-pyridyl isotiocyanate, acmc-20an1z, 3-pyridyl-isothiocyanate, pyridin-3-isothiocyanate
PubChem CID 2737242
IUPAC Name 3-isothiocyanatopyridine
SMILES C1=CC(=CN=C1)N=C=S

Specifications

CAS Min % 94.5
CAS Max % 100.0
Density 1.22g/mL
Assay Percent Range 95%
Packaging Amber Glass Bottle
Quantity 1 g
Formula Weight 136.18
Percent Purity 95%
Chemical Name or Material 3-Pyridyl isothiocyanate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.