missing translation for 'onlineSavingsMsg'
Learn More

3-Pyridyloxyacetic acid, 98%

Product Code. 11456516
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
Unit Size:
1g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11456516 1 g 1g
11466516 5 g 5g
11476516 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11456516 Supplier Thermo Scientific Alfa Aesar Supplier No. B24780.03
 Restricted Item
Estimated Shipment: 18-08-2026
Add to basket
This item is not returnable. View return policy

CAS: 86649-57-2 | C7H7NO3 | 153.14 g/mol

3-Pyridyloxyacetic acid is used in synthesis of raw material for fine chemicals, organic intermediate, pharmaceutical intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
3-Pyridyloxyacetic acid is used in synthesis of raw material for fine chemicals, organic intermediate, pharmaceutical intermediate.

Solubility
Slightly Soluble in water (2.0 g/L)

Notes
Keep container tightly closed. Store away from oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 86649-57-2
Molecular Formula C7H7NO3
Molecular Weight (g/mol) 153.14
MDL Number MFCD04039772
InChI Key MBEPWLCDXKKANL-UHFFFAOYSA-N
Synonym pyridin-3-yloxy-acetic acid, 2-pyridin-3-yloxy acetic acid, 3-pyridyloxyacetic acid, pyridin-3-yloxy acetic acid, 2-3-pyridyloxy acetic acid, pyridin-3-yloxy-aceticacid, 2-3-pyridinyloxy acetic acid, 2-pyridin-3-yloxy aceticacid, acetic acid, 3-pyridinyloxy, 2-pyridine-3-yl oxyacetic acid
PubChem CID 3159630
IUPAC Name 2-pyridin-3-yloxyacetic acid
SMILES OC(=O)COC1=CN=CC=C1

Specifications

Melting Point 210°C to 212°C
Quantity 1 g
Solubility Information Slightly Soluble in water (2.0g/L)
Formula Weight 153.14
Percent Purity 98%
Chemical Name or Material 3-Pyridyloxyacetic acid
compliance-icons
Product Identifier
  • 3-Pyridyloxyacetic acid
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/soap.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Supplemental information
  • MIXTURE LIST-Contain: 3-Pyridyloxyacetic acid

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.