missing translation for 'onlineSavingsMsg'
Learn More

3-(Trifluoromethyl)phenyl isocyanate, 98%

Product Code. 11387426
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
Unit Size:
1g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11387426 1 g 1g
11397426 5 g 5g
11307436 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11387426 Supplier Thermo Scientific Alfa Aesar Supplier No. L09448.03
 Restricted Item
In Stock
Add to basket
This item is not returnable. View return policy

CAS: 329-01-1 | C8H4F3NO | 187.12 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 329-01-1
Molecular Formula C8H4F3NO
Molecular Weight (g/mol) 187.12
MDL Number MFCD00002020
InChI Key SXJYSIBLFGQAND-UHFFFAOYSA-N
Synonym 3-trifluoromethyl phenyl isocyanate, 1-isocyanato-3-trifluoromethyl benzene, benzene, 1-isocyanato-3-trifluoromethyl, a,a,a-trifluoro-m-tolyl isocyanate, isocyanobenzotrifluoride, unii-qps7llz6xm, 3-trifluoromethylphenyl isocyanate, alpha,alpha,alpha-trifluoro-m-tolyl isocyanate, 3-trifluoromethyl phenylisocyanate, qps7llz6xm
PubChem CID 9483
IUPAC Name 1-isocyanato-3-(trifluoromethyl)benzene
SMILES FC(F)(F)C1=CC=CC(=C1)N=C=O

Specifications

Density 1.336
Boiling Point 53°C to 54°C (11 mmHg)
Flash Point 58°C (136°F)
Odor Pungent
Refractive Index 1.4685
Quantity 1 g
UN Number UN2285
Beilstein 744880
Sensitivity Moisture sensitive
Formula Weight 187.12
Percent Purity 98%
Chemical Name or Material 3-(Trifluoromethyl)phenyl isocyanate
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.