missing translation for 'onlineSavingsMsg'
Learn More

(3Beta,5Alpha)-4,4-Dimethylcholesta-8,14,24-trien-3-ol-d6 (Contain up to 15% Delta25 isomer), TRC

Product Code. 30377105
Change view
Click to view available options
Quantity:
0.25 mg
1 mg
5 mg
Unit Size:
0.25mg
1mg
5mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30377105 1 mg 1mg
30376858 0.25 mg 0.25mg
30368882 5 mg 5mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30377105 Supplier Toronto Research Chemicals Supplier No. TRCD4717271MG
 Restricted Item
Estimated Shipment: 29-09-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material (3beta,5alpha)-4,4-Dimethylcholesta-8,14,24-trien-3-ol-d6
Molecular Formula C29H40D6O
Quantity 1 mg
Synonym 4,4-Dimethyl-5α-cholesta-8,14,24-trien-3β-ol-d6, FF-MAS-d6, Follicular Fluid Meiosis-activating Sterol-d6
InChI Formula InChI=1S/C29H46O/c1-19(2)9-8-10-20(3)22-12-13-23-21-11-14-25-27(4,5)26(30)16-18-29(25,7)24(21)15-17-28(22,23)6/h9,13,20,22,25-26,30H,8,10-12,14-18H2,1-7H3/t20-,22-,25+,26+,28-,29-/m1/s1/i4D3,5D3
SMILES [H][C@]1([C@H](C)CC/C=C(C)\C)CC=C([C@]1(C)CC2)C3=C2[C@]4(C)[C@](CC3)([H])C(C([2H])([2H])[2H])(C([2H])([2H])[2H])[C@@H](O)CC4
Molecular Weight (g/mol) 416.71
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.