Learn More
4-(2-Pyridyl)-2-methyl-3-butyn-2-ol, 98+%
Product Code. 11328626
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11328626 1 g 1g
11338626 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 11328626 Supplier Thermo Scientific Alfa Aesar Supplier No. L17676.03
Request Bulk or Custom Format
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
CAS: 29767-97-3 | C10H11NO | 161.204 g/mol
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 29767-97-3
Molecular Formula C10H11NO
Molecular Weight (g/mol) 161.204
MDL Number MFCD00168867
InChI Key YBUWIVMOMHEALF-UHFFFAOYSA-N
Synonym 2-methyl-4-pyridin-2-yl but-3-yn-2-ol, 4-2-pyridyl-2-methyl-3-butyn-2-ol, 2-methyl-4-2-pyridinyl-3-butyn-2-ol, 2-methyl-4-2-pyridyl but-3-yn-2-ol, acmc-20amxd, cambridge id 5119550, 3-methyl-3-hydroxybutyn-1-yl pyridine, 2-methyl-4-2-pyridyl-3-butyn-2-ol, 3-butyn-2-ol, 2-methyl-4-2-pyridinyl
PubChem CID 698810
IUPAC Name 2-methyl-4-pyridin-2-ylbut-3-yn-2-ol
SMILES CC(C)(C#CC1=CC=CC=N1)O
Specifications
Melting Point 80°C to 85°C
Boiling Point 120°C (1.5 mmHg)
Quantity 1 g
Beilstein 1447913
Formula Weight 161.2
Percent Purity ≥98%
Chemical Name or Material 4-(2-Pyridyl)-2-methyl-3-butyn-2-ol