missing translation for 'onlineSavingsMsg'
Learn More

4-(3',6'-Dimethyl-3'-heptyl)phenol Monoethoxylate-13C6, TRC

Product Code. 17951805
Change view
Click to view available options
Quantity:
0.5 mg
1 mg
5 mg
Unit Size:
0.5mg
1mg
5mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
17951805 1 mg 1mg
17994195 0.5 mg 0.5mg
17982715 5 mg 5mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 17951805 Supplier Toronto Research Chemicals Supplier No. TRCD4731171MG
 Restricted Item
Estimated Shipment: 04-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1173019-61-8
Molecular Formula C1113C6H28O2
Molecular Weight (g/mol) 270.36
Synonym 2-[4-(1-Ethyl-1,4-dimethylpentyl)phenoxy]ethanol-13C6
SMILES OCCO[13C]1=[13CH][13CH]=[13C](C(CCC(C)C)(C)CC)[13CH]=[13CH]1

Specifications

Quantity 1 mg
InChI Formula InChI=1S/C17H28O2/c1-5-17(4,11-10-14(2)3)15-6-8-16(9-7-15)19-13-12-18/h6-9,14,18H,5,10-13H2,1-4H3/i6+1,7+1,8+1,9+1,15+1,16+1
Formula Weight 270.2291
Chemical Name or Material 4-(3',6'-Dimethyl-3'-heptyl)phenol Monoethoxylate-13C6
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.