missing translation for 'onlineSavingsMsg'
Learn More

4,4'-Bis(diethylamino)benzophenone, 98+%

Product Code. 11475956
Change view
Click to view available options
Quantity:
25 g
100 g
Unit Size:
100g
25g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11475956 25 g 25g
11485956 100 g 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11475956 Supplier Thermo Scientific Alfa Aesar Supplier No. B22288.14
Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 90-93-7 | C21H28N2O | 324.47 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Identificadores de productos químicos

CAS 90-93-7
Molecular Formula C21H28N2O
Molecular Weight (g/mol) 324.47
MDL Number MFCD00009044
InChI Key VYHBFRJRBHMIQZ-UHFFFAOYSA-N
Synonym 4,4'-bis diethylamino benzophenone, michler's ethyl ketone, bis 4-diethylamino phenyl methanone, methanone, bis 4-diethylamino phenyl, 4,4'-tetraethyldiamino benzophenone, p,p'-bis diethylamino benzophenone, p,p'-tetraethyldiamino benzophenone, ethyl michler ketone
PubChem CID 66663
IUPAC Name bis[4-(diethylamino)phenyl]methanone
SMILES CCN(CC)C1=CC=C(C=C1)C(=O)C1=CC=C(C=C1)N(CC)CC

Especificaciones

Melting Point 93°C to 95°C
pH 4 to 6
Flash Point 151°C (303°F)
Odor Odorless
Quantity 25 g
UN Number UN3077
Beilstein 2148611
Formula Weight 324.47
Percent Purity 99%
Chemical Name or Material 4,4'-Bis(diethylamino)benzophenone

RUO – Research Use Only

Título del producto
Seleccione un problema

Al hacer clic en Enviar, acepta que Fisher Scientific se ponga en contacto con usted en relación con los comentarios que ha proporcionado en este formulario. No compartiremos su información para ningún otro fin. Toda la información de contacto proporcionada se mantendrá de acuerdo con nuestra Política de Privacidad. Política de privacidad.