missing translation for 'onlineSavingsMsg'
Learn More

4-(4-Bromo-1H-pyrazol-1-yl)benzaldehyde, 95%, Thermo Scientific™

Product Code. 10139629
Change view
Click to view available options
Quantity:
1 g
Unit Size:
1g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
10139629 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 10139629 Supplier Thermo Scientific Maybridge Supplier No. MAY00329DA
Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Specifications

CAS 1174064-63-1
Assay Percent Range 95%
Molecular Formula C10H7BrN2O
Quantity 1 g
Synonym 4-4-bromo-1h-pyrazol-1-yl benzaldehyde, 4-4-bromopyrazol-1-yl benzaldehyde, 4-4-bromopyrazolyl benzaldehyde
InChI Key WRORMQDKQSKVFB-UHFFFAOYSA-N
SMILES C1=CC(=CC=C1C=O)N2C=C(C=N2)Br
IUPAC Name 4-(4-bromopyrazol-1-yl)benzaldehyde
Molecular Weight (g/mol) 251.083
PubChem CID 45594325
Formula Weight 251.08
Chemical Name or Material 4-(4-Bromo-1H-pyrazol-1-yl)benzaldehyde
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.