Learn More
4-(4-Nitrophenyl)morpholine, 98%, Thermo Scientific™
Product Code. 11491437
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
Unit Size:
100g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11491437 100 g 100g
11471437 5 g 5g
11481437 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Product Code. 11491437 Supplier Thermo Scientific Alfa Aesar Supplier No. B24933.22
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
missing translation for 'validMainframeMsg'
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 10389-51-2
Molecular Formula C10H12N2O3
Molecular Weight (g/mol) 208.217
MDL Number MFCD00023317
InChI Key IAJDSUYFELYZCS-UHFFFAOYSA-N
Synonym 4-4-nitrophenyl morpholine, morpholine, 4-4-nitrophenyl, p-morpholinonitrobenzene, 4-p-nitrophenyl morpholine, 4-morpholinyl nitrobenzene, 4-morpholinonitrobenzene, n-p-nitrophenyl-morpholine, morpholine, 4-p-nitrophenyl, n-4-nitrophenyl morpholine
PubChem CID 25222
IUPAC Name 4-(4-nitrophenyl)morpholine
SMILES C1COCCN1C2=CC=C(C=C2)[N+](=O)[O-]
Specifications
Melting Point 148°C to 152°C
Quantity 100 g
Formula Weight 208.22
Percent Purity 98%
Chemical Name or Material 4-(4-Nitrophenyl)morpholine