missing translation for 'onlineSavingsMsg'
Learn More

4-(6-Methoxy-3-pyridinyl)-1-piperazinecarboxylic Acid-d4 1,1-Dimethylethyl Ester, TRC

Product Code. 30482991
Change view
Click to view available options
Quantity:
1 mg
2.5 mg
5 mg
Unit Size:
1mg
2.5mg
5mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30482991 1 mg 1mg
30468537 5 mg 5mg
30470615 2.5 mg 2.5mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Product Code. 30482991 Supplier Toronto Research Chemicals Supplier No. TRCM2718071MG
 Restricted Item
Estimated Shipment: 10-09-2026
Add to basket
Add to basket
This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material 4-(6-Methoxy-3-pyridinyl)-1-piperazinecarboxylic Acid-d4 1,1-Dimethylethyl Ester
Molecular Formula C9 H15 N O4
Quantity 1 mg
InChI Formula InChI=1S/C9H15NO4/c10-8(7(11)12)1-3-9(4-2-8)13-5-6-14-9/h1-6,10H2,(H,11,12)
SMILES NC1(CCC2(CC1)OCCO2)C(=O)O
IUPAC Name 8-amino-1,4-dioxaspiro[4.5]decane-8-carboxylic acid
Molecular Weight (g/mol) 201.22
Formula Weight 201.1
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.