missing translation for 'onlineSavingsMsg'
Learn More

4-Acetylbenzenesulfonyl chloride, 97%, Thermo Scientific Chemicals

Product Code. 11395359
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
This item is not returnable. View return policy

Product Code. 11395359

Brand: Thermo Scientific Alfa Aesar H50380.03

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on 056 260 260 for assistance.
View alternative products

This item is not returnable. View return policy

4-Acetylbenzenesulfonyl chloride is used as a sulfanilamide drug intermediary

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
4-Acetylbenzenesulfonyl chloride is used as a sulfanilamide drug intermediary

Solubility
Soluble in water (201.8 mg/L) 25 deg C.

Notes
Moisture Sensitive. Keep container tightly closed. Store in cool, dry conditions in well sealed containers.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1788-10-9
Molecular Formula C8H7ClO3S
Molecular Weight (g/mol) 218.651
MDL Number MFCD00800269
InChI Key FXVDNCRTKXMSEZ-UHFFFAOYSA-N
Synonym 4-acetylbenzene-1-sulfonyl chloride, 4-acetyl-benzenesulfonyl chloride, 4-acetylphenylsulfonyl chloride, benzenesulfonylchloride, 4-acetyl, benzenesulfonyl chloride, 4-acetyl, 1-acetyl-4-chlorosulfonyl benzene, 4-acetylbenzenesulfonyl chloride, acmc-1bs75, ksc183k3b, 4-acetylbenzenesulfonylchloride
PubChem CID 266418
IUPAC Name 4-acetylbenzenesulfonyl chloride
SMILES CC(=O)C1=CC=C(C=C1)S(=O)(=O)Cl

Specifications

Melting Point 84°C
Quantity 1 g
UN Number UN3261
Formula Weight 218.66
Percent Purity ≥99%
Chemical Name or Material 4-Acetylbenzenesulfonyl chloride

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.