missing translation for 'onlineSavingsMsg'
Learn More

4-Acetyldiphenyl sulfone, 98%

Product Code. 11488297
Click to view available options
Quantity:
1 g
5 g
25 g
Unit Size:
1g
25g
5g
This item is not returnable. View return policy

Product Code. 11488297

Brand: Thermo Scientific Alfa Aesar A18376.03

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 65085-83-8 | C14H12O3S | 260.307 g/mol

4-Acetyldiphenyl sulfone is used as an intermediate in organic synthesis.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
4-Acetyldiphenyl sulfone is used as an intermediate in organic synthesis.

Notes
Incompatible with strong oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 65085-83-8
Molecular Formula C14H12O3S
Molecular Weight (g/mol) 260.307
MDL Number MFCD00209628
InChI Key FGFHDNIGKVTTLC-UHFFFAOYSA-N
Synonym 4-acetyldiphenyl sulfone, 1-4-phenylsulfonyl phenyl ethanone, 1-4-benzenesulfonyl phenyl ethanone, 4-acetyl diphenyl sulfone, 1-4-phenylsulfonyl phenyl ethan-1-one, pubchem15334, 4-acetyldiphenylsulfone, 4-phenylsulfonyl acetophenone, phenyl 4-acetylphenyl sulfone
PubChem CID 265878
IUPAC Name 1-[4-(benzenesulfonyl)phenyl]ethanone
SMILES CC(=O)C1=CC=C(C=C1)S(=O)(=O)C2=CC=CC=C2

Specifications

Melting Point 138°C to 140°C
Quantity 1 g
Formula Weight 260.31
Percent Purity 98%
Chemical Name or Material 4-Acetyldiphenyl sulfone

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.