missing translation for 'onlineSavingsMsg'
Learn More

4-Acryloylmorpholine, 98+%, stab. with 4-methoxyphenol

Product Code. 11307816
Change view
Click to view available options
Quantity:
5 g
25 g
Unit Size:
25g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11307816 5 g 5g
11317816 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11307816 Supplier Thermo Scientific Alfa Aesar Supplier No. L12947.06
 Restricted Item
Estimated Shipment: 12-08-2026
Add to basket
This item is not returnable. View return policy

CAS: 5117-12-4 | C7H11NO2 | 141.17 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 5117-12-4
Molecular Formula C7H11NO2
Molecular Weight (g/mol) 141.17
MDL Number MFCD00047413
InChI Key XLPJNCYCZORXHG-UHFFFAOYSA-N
Synonym 4-acryloylmorpholine, 1-morpholinoprop-2-en-1-one, n-acrylylmorpholine, n-acryloylmorpholine, morpholine, 4-1-oxo-2-propenyl, poly acryloylmorpholine, acryloylmorpholine, acryloyl morpholine, 1-morpholin-4-yl prop-2-en-1-one, unii-k0y58p61ja
PubChem CID 98723
IUPAC Name 1-morpholin-4-ylprop-2-en-1-one
SMILES C=CC(=O)N1CCOCC1

Specifications

Melting Point -8°C
Density 1.12
Boiling Point 158°C (50 mmHg)
Flash Point 127°C (260°F)
Refractive Index 1.509
Quantity 5 g
Beilstein 119302
Sensitivity Light sensitive
Formula Weight 141.17
Percent Purity ≥98%
Chemical Name or Material 4-Acryloylmorpholine, stabilized with 4-methoxyphenol
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.