missing translation for 'onlineSavingsMsg'
Learn More

4'-Benzyloxy-2'-hydroxyacetophenone, 98%, Thermo Scientific™

Product Code. 15555946 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
1 g
Unit Size:
1g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15555946 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 15555946 Supplier Thermo Scientific Acros Supplier No. 457130010
Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Specifications

CAS 29682-12-0
Melting Point 104.0°C to 106.0°C
Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (HPLC)
Packaging Glass Bottle
Molecular Formula C15H14O3
Quantity 1 g
Synonym 1-4-benzyloxy-2-hydroxyphenyl ethanone, 4'-benzyloxy-2'-hydroxyacetophenone, 4-benzyloxy-2-hydroxyacetophenone, 2-hydroxy-4-benzyloxyacetophenone, ethanone, 1-2-hydroxy-4-phenylmethoxy phenyl, 1-2-hydroxy-4-phenylmethoxyphenyl ethanone, 1-2-hydroxy-4-phenylmethoxy-phenyl ethanone, 1-4-benzyloxy-2-hydroxyphenyl ethan-1-one
InChI Key AGQNLHOTLJFJCG-UHFFFAOYSA-N
SMILES CC(=O)C1=C(C=C(C=C1)OCC2=CC=CC=C2)O
IUPAC Name 1-(2-hydroxy-4-phenylmethoxyphenyl)ethanone
Molecular Weight (g/mol) 242.28
PubChem CID 309257
Formula Weight 242.28
Percent Purity 98%
Chemical Name or Material 4'-Benzyloxy-2'-hydroxyacetophenone
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.