missing translation for 'onlineSavingsMsg'
Learn More

4-(Benzyloxycarbamoyl)benzeneboronic acid pinacol ester, 95%, Thermo Scientific™

Product Code. 15429705
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
Product Code. Quantity unitSize
15429705 1 g 1g
15439705 5 g 5g
2 options
This item is not returnable. View return policy

Product Code. 15429705

Brand: Thermo Scientific Alfa Aesar H52282.03

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on 056 260 260 for assistance.
View alternative products

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 2096330-26-4
Molecular Formula C20H24BNO4
Molecular Weight (g/mol) 353.23
MDL Number MFCD18827484
InChI Key XECXLZBBBRYLFW-UHFFFAOYSA-N
Synonym n-benzyloxy-4-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl benzamide, n-benzyloxy-4-4,4,5,5-tetramethyl-1,3,2 dioxaborolan-2-yl benzamide, amtb379, 4-benzyloxycarbamoyl benzeneboronic acid pinacol ester, n-benzyloxy-4-4,4,5,5-tetramethyl-1,3,2 dioxaborolan-2-yl-benzamide
PubChem CID 71306540

Specifications

Quantity 1 g
Formula Weight 353.22
Percent Purity 95%
Chemical Name or Material 4-(Benzyloxycarbamoyl)benzeneboronic acid pinacol ester
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.