missing translation for 'onlineSavingsMsg'
Learn More

4-Chloro-1-methylpiperidine hydrochloride, 97%

Product Code. 11327555
Change view
Click to view available options
Quantity:
25 g
100 g
Unit Size:
100g
25g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11327555 25 g 25g
11337555 100 g 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11327555 Supplier Thermo Scientific Alfa Aesar Supplier No. L05699.14
 Restricted Item
Estimated Shipment: 17-08-2026
Add to basket
This item is not returnable. View return policy

CAS: 5382-23-0 | C6H13Cl2N | 170.077 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 5382-23-0
Molecular Formula C6H13Cl2N
Molecular Weight (g/mol) 170.077
MDL Number MFCD00012834
InChI Key QDOJNWFOCOWAPO-UHFFFAOYSA-N
Synonym 4-chloro-1-methylpiperidine hydrochloride, 1-methyl-4-chloropiperidine hydrochloride, 4-chloro-n-methylpiperidine hydrochloride, 4-chloro-1-methylpiperidinium chloride, piperidine, 4-chloro-1-methyl-, hydrochloride, n-methyl-4-chloropiperidine hydrochloride, 1-methyl-4-chloro piperidine, pubchem2461, acmc-1apdo, ksc495i8n
PubChem CID 3034158
IUPAC Name 4-chloro-1-methylpiperidine;hydrochloride
SMILES CN1CCC(CC1)Cl.Cl

Specifications

Melting Point 159°C to 166°C
Quantity 25 g
Beilstein 3677071
Sensitivity Hygroscopic
Formula Weight 170.08
Percent Purity 0.97
Chemical Name or Material 4-Chloro-1-methylpiperidine hydrochloride

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.