missing translation for 'onlineSavingsMsg'
Learn More

4-Chlorobenzeneboronic acid, 98+%

Product Code. 11447577
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
Unit Size:
1g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11447577 1 g 1g
11457577 5 g 5g
11467577 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11447577 Supplier Thermo Scientific Alfa Aesar Supplier No. A15657.03
 Restricted Item
Estimated Shipment: 18-08-2026
Add to basket
This item is not returnable. View return policy

CAS: 1679-18-1 | C6H6BClO2 | 156.37 g/mol

Used as a GC reagent for diols. It is a reagent used for palladium-catalyzed direct arylation, cyclopalladation, tandem-type Pd(II)-catalyzed oxidative Heck reaction and intramolecular C-H amidation, copper-mediated ligandless aerobic fluoroalkylation, Pd-catalyzed arylative cyclization, ruthenium catalyzed direct arylation, ligand-free copper-catalyzed coupling reactions and regioselective arylation and alkynylation by Suzuki-Miyaura and Sonogashira cross-coupling reactions. Also in as a reagent in the preparation of catalysts for Suzuki-Miyaura cross-coupling, palladium(II) thiocarboxamide complexes as Suzuki coupling catalysts, Biaryls by Suzuki reactions of aryl chlorides, bromides, and iodides with arylboronic acids.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Used as a GC reagent for diols. It is a reagent used for palladium-catalyzed direct arylation, cyclopalladation, tandem-type Pd(II)-catalyzed oxidative Heck reaction and intramolecular C-H amidation, copper-mediated ligandless aerobic fluoroalkylation, Pd-catalyzed arylative cyclization, ruthenium catalyzed direct arylation, ligand-free copper-catalyzed coupling reactions and regioselective arylation and alkynylation by Suzuki-Miyaura and Sonogashira cross-coupling reactions. Also in as a reagent in the preparation of catalysts for Suzuki-Miyaura cross-coupling, palladium(II) thiocarboxamide complexes as Suzuki coupling catalysts, Biaryls by Suzuki reactions of aryl chlorides, bromides, and iodides with arylboronic acids.

Solubility
Slightly soluble in water. Soluble in DMSO.

Notes
Store away from oxidizing agents. Keep the container tightly closed and place it in a cool, dry and well ventilated condition.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1679-18-1
Molecular Formula C6H6BClO2
Molecular Weight (g/mol) 156.37
MDL Number MFCD00039137
InChI Key CAYQIZIAYYNFCS-UHFFFAOYSA-N
Synonym 4-chlorophenyl boronic acid, 4-chlorobenzeneboronic acid, p-chlorophenylboronic acid, benzeneboronic acid, p-chloro, p-chlorobenzeneboronic acid, boronic acid, 4-chlorophenyl, boronic acid, p-chlorophenyl, 4-chlorophenyl boranediol, 4-chlorophenylbornic acid
PubChem CID 74299
IUPAC Name (4-chlorophenyl)boronic acid
SMILES OB(O)C1=CC=C(Cl)C=C1

Specifications

Melting Point 268°C to 277°C
Quantity 1 g
Beilstein 2936346
Solubility Information Slightly soluble in water. Soluble in DMSO.
Formula Weight 156.38
Percent Purity ≥98%
Chemical Name or Material 4-Chlorobenzeneboronic acid

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.