missing translation for 'onlineSavingsMsg'
Learn More

4-Chlorobenzhydrol, TRC

Product Code. 30136415
Change view
Click to view available options
Quantity:
1 g
10 g
50 g
Unit Size:
10g
1g
50g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30136415 50 g 50g
30134375 10 g 10g
17970425 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30136415 Supplier Toronto Research Chemicals Supplier No. TRCC36449050G
 Restricted Item
Estimated Shipment: 09-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 119-56-2
Molecular Formula C13 H11 Cl O
Molecular Weight (g/mol) 218.68
Synonym (RS)-(4-Chlorophenyl)phenylmethanol, 4-Chlorobenzhydrol, Meclozine Hydrochloride Imp. B (EP)
IUPAC Name (4-chlorophenyl)-phenylmethanol
SMILES OC(c1ccccc1)c2ccc(Cl)cc2

Specifications

Quantity 50 g
InChI Formula InChI=1S/C13H11ClO/c14-12-8-6-11(7-9-12)13(15)10-4-2-1-3-5-10/h1-9,13,15H
Formula Weight 218.0498
Percent Purity >95%
Chemical Name or Material 4-Chlorobenzhydrol
Grade HPLC
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.