missing translation for 'onlineSavingsMsg'
Learn More

4-Fluoro-2-methoxypyridine, 97%, Thermo Scientific™

Product Code. 15512313 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
1 g
Unit Size:
1g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15512313 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 15512313 Supplier Thermo Scientific Acros Supplier No. 450010010
 Restricted Item
Estimated Shipment: 03-12-2026
Add to basket
This item is not returnable. View return policy

Specifications

CAS 96530-81-3
Infrared Spectrum Authentic
Assay Percent Range 96% min. (HPLC)
Packaging Glass Bottle
Molecular Formula C6H6FNO
MDL Number MFCD09835267
Quantity 1 g
Synonym pyridine, 4-fluoro-2-methoxy, 4-fluoro-2-methoxy-pyridine, 4-fluoro 2-methoxy pyridine, 2-methoxy-4-fluoropyridine, fluoromethoxypyridine, pubchem6596, acmc-209v4d, 4-fluoranyl-2-methoxy-pyridine, 3,3'-methylenebis 5-methyloxazolidine, pyridine, 4-fluoro-2-methoxy-9ci
InChI Key RGWFJFBUHTUVMV-UHFFFAOYSA-N
SMILES COC1=NC=CC(F)=C1
IUPAC Name 4-fluoro-2-methoxypyridine
Molecular Weight (g/mol) 127.12
PubChem CID 338458
Formula Weight 127.12
Percent Purity 97%
Chemical Name or Material 4-Fluoro-2-methoxypyridine
Show More Show Less
compliance-icons
Product Identifier
  • 4-Fluoro-2-methoxypyridine
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H312-Harmful in contact with skin.
  • H332-Harmful if inhaled.
  • H302-Harmful if swallowed.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.