Learn More
4-Hydroxy-3,5-dimethylbenzaldehyde, 97+%
Product Code. 11416167
Click to view available options
Quantity:
1 g
5 g
25 g
Unit Size:
1g
25g
5g
Product Code. 11416167
Brand: Thermo Scientific Alfa Aesar B25123.03
Request Bulk or Custom Format
CAS: 2233-18-3 | C9H10O2 | 150.18 g/mol
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 2233-18-3
Molecular Formula C9H10O2
Molecular Weight (g/mol) 150.18
MDL Number MFCD00006946
InChI Key UYGBSRJODQHNLQ-UHFFFAOYSA-N
Synonym 3,5-dimethyl-4-hydroxybenzaldehyde, benzaldehyde, 4-hydroxy-3,5-dimethyl, 4-formyl-2,6-xylenol, 4-hydroxy-3,5-dimethyl-benzaldehyde, unii-23pa2pmp9f, 23pa2pmp9f, syringnldehyde, zlchem 510, pubchem8252, buttpark 9657-68
PubChem CID 75222
IUPAC Name 4-hydroxy-3,5-dimethylbenzaldehyde
SMILES CC1=CC(C=O)=CC(C)=C1O
Specifications
Melting Point 112°C to 116°C
Quantity 1 g
Beilstein 1908717
Sensitivity Air Sensitive
Formula Weight 150.18
Percent Purity 98%
Chemical Name or Material 4-Hydroxy-3,5-dimethylbenzaldehyde