missing translation for 'onlineSavingsMsg'
Learn More

4-Hydroxy-isophthalic Acid Dimethyl Ester, TRC

Product Code. 30135472
Change view
Click to view available options
Quantity:
50 mg
500 mg
1 g
Unit Size:
1g
500mg
50mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30135472 500 mg 500mg
30135220 1 g 1g
30004178 50 mg 50mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30135472 Supplier Toronto Research Chemicals Supplier No. TRCH953760500MG
 Restricted Item
Estimated Shipment: 31-07-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 5985-24-0
Molecular Formula C10H10O5
Molecular Weight (g/mol) 210.18
Synonym 1,3-Benzenedicarboxylic acid, 4-hydroxy-, 1,3-dimethyl ester, 1,3-Benzenedicarboxylic acid, 4-hydroxy-, dimethyl ester (9CI), Isophthalic acid, 4-hydroxy-, dimethyl ester (6CI,7CI,8CI), 4-Hydroxyisophthalic acid dimethyl ester, Dimethyl 4-hydroxyisophthalate, NSC 109108, Dimethyl 4-hydroxyisophthalate, Dimethyl 4-Hydroxybenzene-1,3-dicarboxylate
IUPAC Name dimethyl 4-hydroxybenzene-1,3-dicarboxylate
SMILES COC(=O)c1ccc(O)c(c1)C(=O)OC

Specifications

Quantity 500 mg
InChI Formula InChI=1S/C10H10O5/c1-14-9(12)6-3-4-8(11)7(5-6)10(13)15-2/h3-5,11H,1-2H3
Formula Weight 210.0528
Chemical Name or Material Dimethyl 4-hydroxyisophthalate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.