missing translation for 'onlineSavingsMsg'
Learn More

4-Hydroxyphenylarsonic Acid, TRC

Product Code. 30399728
Change view
Click to view available options
Quantity:
250 mg
500 mg
2.5 g
Unit Size:
2.5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30399728 250 mg -
30408346 500 mg -
30397734 2.5 g 2.5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30399728 Supplier Toronto Research Chemicals Supplier No. TRCH999980250MG
 Restricted Item

This item requires a price update prior to being ordered.

Request a Price for this item here.

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 98-14-6
Molecular Formula C6H7O4As
Molecular Weight (g/mol) 218.03
IUPAC Name (4-hydroxyphenyl)arsonic acid
SMILES Oc1ccc(cc1)[As](=O)(O)O

Specifications

Quantity 250 mg
InChI Formula InChI=1S/C6H7AsO4/c8-6-3-1-5(2-4-6)7(9,10)11/h1-4,8H,(H2,9,10,11)
Formula Weight 217.956
Chemical Name or Material 4-Hydroxyphenylarsonic Acid
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.