missing translation for 'onlineSavingsMsg'
Learn More

4-Methylumbelliferyl Sulfate Potassium Salt, TRC

Product Code. 30006503
Change view
Click to view available options
Quantity:
500 mg
1 g
Unit Size:
1g
500mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30006503 1 g 1g
30006721 500 mg 500mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30006503 Supplier Toronto Research Chemicals Supplier No. TRCM3331001G
 Restricted Item
Estimated Shipment: 25-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 15220-11-8
Molecular Formula C10 H7 O6 S . K
Molecular Weight (g/mol) 294.32
IUPAC Name potassium;(4-methyl-2-oxochromen-7-yl) sulfate
SMILES [K+].CC1=CC(=O)Oc2cc(OS(=O)(=O)[O-])ccc12

Specifications

Quantity 1 g
InChI Formula InChI=1S/C10H8O6S.K/c1-6-4-10(11)15-9-5-7(2-3-8(6)9)16-17(12,13)14;/h2-5H,1H3,(H,12,13,14);/q;+1/p-1
Formula Weight 293.96
Chemical Name or Material 4-Methylumbelliferyl Sulfate, Potassium Salt
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.