missing translation for 'onlineSavingsMsg'
Learn More

4'-n-Butylacetophenone, 97%

Product Code. 11408166
Change view
Click to view available options
Quantity:
5 g
25 g
Unit Size:
25g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11408166 5 g 5g
11418166 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11408166 Supplier Thermo Scientific Alfa Aesar Supplier No. A15296.06
 Restricted Item
Estimated Shipment: 24-08-2026
Add to basket
This item is not returnable. View return policy

CAS: 37920-25-5 | C12H16O | 176.26 g/mol

4'-n-Butylacetophenone is used as an organic chemical synthesis intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
4′-n-Butylacetophenone is used as an organic chemical synthesis intermediate.

Solubility
Not miscible in water.

Notes
Stable under recommended storage conditions. Incompatible with oxidizing agents, acids and bases.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 37920-25-5
Molecular Formula C12H16O
Molecular Weight (g/mol) 176.26
MDL Number MFCD00017500
InChI Key MQESVSITPLILCO-UHFFFAOYSA-N
Synonym 4'-butylacetophenone, 1-4-butylphenyl ethanone, ethanone, 1-4-butylphenyl, p-butylacetophenone, 1-4-butylphenyl ethan-1-one, p-n-butylacetophenone, 4'-n-butylacetophenone, unii-6738cf9z8v, 1-acetyl-4-butylbenzene, 1-4-butyl-phenyl ethanone
PubChem CID 123470
IUPAC Name 1-(4-butylphenyl)ethanone
SMILES CCCCC1=CC=C(C=C1)C(C)=O

Spezifikation

Density 0.957
Boiling Point 101°C to 102°C (1.5 mmHg)
Flash Point >110°C (230°F)
Quantity 5 g
Solubility Information Not miscible in water.
Formula Weight 176.26
Percent Purity 97%
Chemical Name or Material 4'-n-Butylacetophenone

RUO – Research Use Only

Name des Produkts
Wählen Sie ein Thema

Indem Sie auf Absenden klicken, erklären Sie sich damit einverstanden, dass Fisher Scientific sich mit Ihnen in Verbindung setzen kann, um Ihr Feedback in diesem Formular zu bearbeiten. Wir werden Ihre Informationen nicht für andere Zwecke weitergeben. Alle bereitgestellten Kontaktinformationen werden in Übereinstimmung mit unserer Datenschutzrichtlinie aufbewahrt. Datenschutzrichtlinie.