Learn More
4-(N-tert-Butoxycarbonyl)aminobenzoic Acid-d4, TRC
Specifications
Specifications
| Chemical Name or Material | 4-(N-tert-Butoxycarbonyl)aminobenzoic Acid-d4 |
| Molecular Formula | C12 D4 H11 N O4 |
| Quantity | 1 mg |
| Synonym | 4-(tert-Butoxycarbonylamino)benzoic Acid-d4, p-(tert-Butoxycarbonylamino)benzoic Acid-d4, (4-Carboxyphenyl)carbamic Acid tert-Butyl Ester-d4, 4-[[(1,1-Dimethylethoxy)carbonyl]amino]benzoic Acid-d4 |
| InChI Formula | InChI=1S/C12H15NO4/c1-12(2,3)17-11(16)13-9-6-4-8(5-7-9)10(14)15/h4-7H,1-3H3,(H,13,16)(H,14,15)/i4D,5D,6D,7D |
| SMILES | [2H]c1c([2H])c(C(=O)O)c([2H])c([2H])c1NC(=O)OC(C)(C)C |
| IUPAC Name | 2,3,5,6-tetradeuterio-4-[(2-methylpropan-2-yl)oxycarbonylamino]benzoic acid |
| Molecular Weight (g/mol) | 241.276 |
| Formula Weight | 241.125 |