missing translation for 'onlineSavingsMsg'
Learn More

4-Nitropyrene, TRC

Product Code. 30412553
Change view
Click to view available options
Quantity:
25 mg
100 mg
250 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity
30412553 250 mg
30421954 100 mg
30418599 25 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30412553 Supplier Toronto Research Chemicals Supplier No. TRCN900850250MG
 Restricted Item
Estimated Shipment: 04-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 57835-92-4
Molecular Formula C16 H9 N O2
Molecular Weight (g/mol) 247.25
Synonym Pyrene, 4-nitro-, 4-Nitropyrene
IUPAC Name 4-nitropyrene
SMILES [O-][N+](=O)c1cc2cccc3ccc4cccc1c4c23

Specifications

Quantity 250 mg
InChI Formula InChI=1S/C16H9NO2/c18-17(19)14-9-12-5-1-3-10-7-8-11-4-2-6-13(14)16(11)15(10)12/h1-9H
Formula Weight 247.0633
Chemical Name or Material 4-Nitropyrene
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.