missing translation for 'onlineSavingsMsg'
Learn More

4-Phthalimido-2-butyne, 97%, Thermo Scientific™

Product Code. 10310684 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
1 g
5 g
Packaging:
Glass bottle
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
10310684 5 g Glass bottle 5g
10519813 1 g Glass bottle 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10310684 Supplier Thermo Scientific Acros Supplier No. 382150050

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Chemical Identifiers

CAS 113439-83-1
Molecular Formula C12H9NO2
Molecular Weight (g/mol) 199.21
MDL Number MFCD00160830
InChI Key COPWPECHFWIYSP-UHFFFAOYSA-N
Synonym n-2-butynyl phthalimide, 2-but-2-yn-1-yl isoindoline-1,3-dione, 2-but-2-yn-1-yl isoindole-1,3-dione, acmc-1bscb, 1-phthalimido-2-butyne, 4-phthalimido-2-butyne, 2-2-butynyl isoindoline-1,3-dione, 1h-isoindole-1,3 2h-dione, 2-2-butyn-1-yl
PubChem CID 8063352
IUPAC Name 2-(but-2-yn-1-yl)-2,3-dihydro-1H-isoindole-1,3-dione
SMILES CC#CCN1C(=O)C2=CC=CC=C2C1=O

Specifications

CAS Min % 96.0
CAS Max % 100.0
Melting Point 216°C to 223°C
Color White to Brown
Infrared Spectrum Authentic
Assay Percent Range 97%
Packaging Glass bottle
Quantity 5 g
Formula Weight 199.21
Percent Purity ≥96.0%
Physical Form Crystalline Powder
Chemical Name or Material 4-Phthalimido-2-butyne
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.