Learn More
4-tert-Butoxybenzonitrile, 96%, Thermo Scientific™
Product Code. 11364895
Change view
Click to view available options
Quantity:
5 g
25 g
Unit Size:
25g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11364895 5 g 5g
11374895 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 11364895 Supplier Thermo Scientific Alfa Aesar Supplier No. L18067.06
Request Bulk or Custom Format
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on 056 260 260 for assistance.
View alternative products
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 185259-36-3
Molecular Formula C11H13NO
Molecular Weight (g/mol) 175.231
MDL Number MFCD03425273
InChI Key WLWYCCYGVDZFMS-UHFFFAOYSA-N
Synonym 4-tert-butoxybenzonitrile, 4-tert-butoxy benzonitrile, benzonitrile, 4-1,1-dimethylethoxy, acmc-20amau, 4-tert-butoxy benzenecarbonitrile, 4-2-methylpropan-2-yl oxy benzonitrile
PubChem CID 7021978
IUPAC Name 4-[(2-methylpropan-2-yl)oxy]benzonitrile
SMILES CC(C)(C)OC1=CC=C(C=C1)C#N
Specifications
Boiling Point 95°C (0.8 mmHg)
Refractive Index 1.521
Quantity 5 g
Beilstein 3237723
Formula Weight 175.23
Percent Purity 96%
Chemical Name or Material 4-tert-Butoxybenzonitrile