missing translation for 'onlineSavingsMsg'
Learn More

4-Trimethylsilyl-3-butyn-1-ol, 98%

Product Code. 15400138
Change view
Click to view available options
Quantity:
5 g
25 g
Unit Size:
25g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15400138 25 g 25g
15490128 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15400138 Supplier Thermo Scientific Alfa Aesar Supplier No. H53376.14
 Restricted Item
Estimated Shipment: 28-09-2026
Add to basket
This item is not returnable. View return policy

CAS: 2117-12-6 | C7H14OSi | 142.27 g/mol

4-Trimethylsilyl-3-butyn-1-ol is used as an organic chemical synthesis intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
4-Trimethylsilyl-3-butyn-1-ol is used as an organic chemical synthesis intermediate.

Notes
Stable under recommended storage conditions. Incompatible with oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 2117-12-6
MDL Number MFCD00190217

Specifications

Boiling Point 97°C to 98°C (24mmHg)
Quantity 25 g
Molecular Formula C10H8N2S2
Linear Formula HOCH2CH2C≡CSi(CH3)3
UN Number UN1993
Flash Point 60°C (140°F)
Synonym 2,2'-dithiodipyridine, 2,2'-dipyridyl disulfide, 1,2-di pyridin-2-yl disulfane, aldrithiol-2, 2,2'-dipyridyldisulfide, aldrithiol 2, bis 2-pyridyl disulfide, 2-pyridin-2-yldisulfanyl pyridine, 2-dipyridyl disulfide, 2-pyridinyl disulfide
InChI Key HAXFWIACAGNFHA-UHFFFAOYSA-N
SMILES C1=CC=NC(=C1)SSC2=CC=CC=N2
IUPAC Name 2-(pyridin-2-yldisulfanyl)pyridine
Molecular Weight (g/mol) 220.31
PubChem CID 65093
Formula Weight 142.27
Percent Purity 98%
Density 0.854
Chemical Name or Material 4-Trimethylsilyl-3-butyn-1-ol
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.