missing translation for 'onlineSavingsMsg'
Learn More

5-Aminolevulinic Acid-3-13C Hydrochloride, TRC

Product Code. 30135437
Change view
Click to view available options
Quantity:
0.5 mg
5 mg
Unit Size:
0.5mg
5mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30135437 0.5 mg 0.5mg
30004474 5 mg 5mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30135437 Supplier Toronto Research Chemicals Supplier No. TRCA6117820.5MG
 Restricted Item
Estimated Shipment: 21-09-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 129720-95-2
Molecular Formula C413CH9NO3•HCl
Molecular Weight (g/mol) 132.123646
Synonym 5-Amino-4-oxopentanoic Acid-13C, Hydrochloride, δ-Aminolevulinic-13C Acid Hydrochloride, Aminolevulinic-13C Acid Hydrochloride, 5-ALA HCl-13C, Levulan-13C;
IUPAC Name 5-amino-4-oxo(313C)pentanoic acid;hydrochloride
SMILES NCC([13CH2]CC(O)=O)=O.Cl

Specifications

Quantity 0.5 mg
InChI Formula InChI=1S/C5H9NO3.ClH/c6-3-4(7)1-2-5(8)9;/h1-3,6H2,(H,8,9);1H/i1+1;
Chemical Name or Material 5-Aminolevulinic Acid-3-13C Hydrochloride
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.