Learn More
5-Iodoisatin, 97%, Thermo Scientific Chemicals
Product Code. 11437207
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
Unit Size:
100g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11437207 5 g 5g
11447207 25 g 25g
11457207 100 g 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Product Code. 11437207 Supplier Thermo Scientific Alfa Aesar Supplier No. A14988.06
 Restricted Item
In Stock
Add to basket
missing translation for 'validMainframeMsg'
CAS: 20780-76-1 | Molecular Formula: C8H4INO2 | Molecular Weight: 273.029 g/mol
5-Iodoisatin undergoes condensation reaction with phenol yields 5-iodophenolisatin, malonic acid yields 6-iodo-2-quinolone-4-carboxylic acid.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
5-Iodoisatin undergoes condensation reaction with phenol yields 5-iodophenolisatin, malonic acid yields 6-iodo-2-quinolone-4-carboxylic acid.
Solubility
Soluble acetone, acetic acid, ethanol. Insoluble in water.
Notes
Light Sensitive. Keep container tightly closed. Store away from oxidizing agents.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 20780-76-1
Molecular Formula C8H4INO2
Molecular Weight (g/mol) 273.029
MDL Number MFCD00016899
InChI Key OEUGDMOJQQLVAZ-UHFFFAOYSA-N
Synonym 5-iodoisatin, 5-iodoindoline-2,3-dione, 5-iodo isatin, 5-iodoindole-2,3-dione, 5-iodo-2,3-dioxoindoline, 1h-indole-2,3-dione, 5-iodo, 5-iodo-2,3-dihydro-1h-indole-2,3-dione, 5-iodo-1h-benzo d azolidine-2,3-dione, 5-iodo-isatine, 5-iodo-isatin
PubChem CID 88695
IUPAC Name 5-iodo-1H-indole-2,3-dione
SMILES C1=CC2=C(C=C1I)C(=O)C(=O)N2
Especificaciones
Melting Point ∼276°C (decomposition)
Quantity 5 g
Beilstein 131248
Sensitivity Light sensitive
Solubility Information Soluble acetone,acetic acid,ethanol. Insoluble in water.
Formula Weight 273.03
Percent Purity 97%
Chemical Name or Material 5-Iodoisatin
RUO – Research Use Only
Título del producto
Seleccione un problema
Al hacer clic en Enviar, acepta que Fisher Scientific se ponga en contacto con usted en relación con los comentarios que ha proporcionado en este formulario. No compartiremos su información para ningún otro fin. Toda la información de contacto proporcionada se mantendrá de acuerdo con nuestra Política de Privacidad. Política de privacidad.