missing translation for 'onlineSavingsMsg'
Learn More

5-Methoxyindole-2-carboxylic acid, 97%

Product Code. 11328166
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11328166 1 g 1g
11338166 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 11328166 Supplier Thermo Scientific Alfa Aesar Supplier No. L15137.03
Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

CAS: 4382-54-1 | C10H9NO3 | 191.19 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 4382-54-1
Molecular Formula C10H9NO3
Molecular Weight (g/mol) 191.19
MDL Number MFCD00005614
InChI Key YEBJVSLNUMZXRJ-UHFFFAOYSA-N
Synonym 5-methoxyindole-2-carboxylic acid, 1h-indole-2-carboxylic acid, 5-methoxy, 5-methoxy-2-indolecarboxylic acid, indole-2-carboxylic acid, 5-methoxy, acide methoxy-5 indole carboxylique-2, acide methoxy-5 indole carboxylique-2 french, 5-methoxyindol-2-carboxylic acid, 5-methoxy-1h-indole-2-carboxylicacid, pubchem1702, acmc-209jvj
PubChem CID 20401
IUPAC Name 5-methoxy-1H-indole-2-carboxylic acid
SMILES COC1=CC=C2NC(=CC2=C1)C(O)=O

Specifications

Melting Point ∽200°C (decomposition)
Quantity 1 g
Beilstein 157478
Formula Weight 191.19
Percent Purity 97%
Chemical Name or Material 5-Methoxyindole-2-carboxylic acid

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.