missing translation for 'onlineSavingsMsg'
Learn More

5-(thien-2-yl)thiophene-2-carbonitrile, Thermo Scientific™

Product Code. 10377634
Change view
Click to view available options
Quantity:
250 mg
1 g
5 g
Unit Size:
1g
250mg
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
10377634 5 g 5g
10447052 250 mg 250mg
10203472 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 10377634 Supplier Thermo Scientific Maybridge Supplier No. SEW02204DE
 Restricted Item
Estimated Shipment: 06-10-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Chemical Identifiers

CAS 16278-99-2
Molecular Formula C9H5NS2
Molecular Weight (g/mol) 191.266
InChI Key ANARYBGZNUMARH-UHFFFAOYSA-N
Synonym 5-thien-2-yl thiophene-2-carbonitrile, 2,2'-bithiophene-5-carbonitrile, 5-thiophen-2-yl thiophene-2-carbonitrile, maybridge3_006613, 5-cyano-2,2'-bithiophene, 5-2-thienyl thiophene-2-carbonitrile
PubChem CID 2739839
IUPAC Name 5-thiophen-2-ylthiophene-2-carbonitrile
SMILES C1=CSC(=C1)C2=CC=C(S2)C#N

Specifications

Assay Percent Range 97%
Packaging Amber glass bottle
Quantity 5 g
Formula Weight 191.28
Percent Purity 97%
Chemical Name or Material 5-(Thien-2-yl)thiophene-2-carbonitrile
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.