Learn More
5-Vinyluracil, TRC
Product Code. 30451929
Change view
Click to view available options
Quantity:
100 mg
250 mg
500 mg
Unit Size:
100mg
250mg
500mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30451929 100 mg 100mg
30455619 250 mg 250mg
30452369 500 mg 500mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Product Code. 30451929 Supplier Toronto Research Chemicals Supplier No. TRCV625000100MG
 Restricted Item
Estimated Shipment: 07-08-2026
Add to basket
Add to basket
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 37107-81-6
Molecular Formula C6 H6 N2 O2
Molecular Weight (g/mol) 138.12
Synonym 2,4(1H,3H)-Pyrimidinedione, 5-ethenyl-, 5-Ethenyl-2,4(1H,3H)-pyrimidinedione, 5-Vinyluracil, NSC 208961
IUPAC Name 5-ethenyl-1H-pyrimidine-2,4-dione
SMILES C=CC1=CNC(=O)NC1=O
Specifications
Quantity 100 mg
InChI Formula InChI=1S/C6H6N2O2/c1-2-4-3-7-6(10)8-5(4)9/h2-3H,1H2,(H2,7,8,9,10)
Formula Weight 138.0429
Chemical Name or Material 5-Vinyluracil