missing translation for 'onlineSavingsMsg'
Learn More

Estradiol Enanthate, TRC

Product Code. 18023700
Change view
Click to view available options
Quantity:
5 g
10 g
25 g
Unit Size:
10g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18023700 5 g 5g
30138255 10 g 10g
30134763 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 18023700 Supplier Toronto Research Chemicals Supplier No. TRCE8881205G

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 4956-37-0
Molecular Formula C25 H36 O3
Molecular Weight (g/mol) 384.55
Synonym Estradiol Enanthate, Estra-1,3,5(10)-triene-3,17-diol (17β)-17-heptanoate, Estradiol-17-enanthate, 3-Hydroxyestra-1,3,5(10)-trien-17β-yl Ester Heptanoic Acid, 17-Heptanoate Estradiol
IUPAC Name [(8R,9S,13S,14S,17S)-3-hydroxy-13-methyl-6,7,8,9,11,12,14,15,16,17-decahydrocyclopenta[a]phenanthren-17-yl] heptanoate
SMILES CCCCCCC(=O)O[C@H]1CC[C@H]2[C@@H]3CCc4cc(O)ccc4[C@H]3CC[C@]12C

Specifications

Quantity 5 g
InChI Formula InChI=1S/C25H36O3/c1-3-4-5-6-7-24(27)28-23-13-12-22-21-10-8-17-16-18(26)9-11-19(17)20(21)14-15-25(22,23)2/h9,11,16,20-23,26H,3-8,10,12-15H2,1-2H3/t20-,21-,22+,23+,25+/m1/s1
Formula Weight 384.2664
Percent Purity >95%
Chemical Name or Material Estradiol Enanthate
Grade HPLC
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.