missing translation for 'onlineSavingsMsg'
Learn More

ent-Kaurenoic Acid, TRC

Product Code. 17908314
Change view
Click to view available options
Quantity:
5 mg
10 mg
25 mg
Unit Size:
10mg
25mg
5mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
17908314 5 mg 5mg
30134535 10 mg 10mg
30134275 25 mg 25mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 17908314 Supplier Toronto Research Chemicals Supplier No. TRCK1456605MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 6730-83-2
Molecular Formula C20H30O2
Molecular Weight (g/mol) 302.45
Synonym (4α)-Kaur-16-en-18-oic Acid, (-)-Kaur-16-en-19-oic Acid, (-)-Kauren-19-oic Acid, Cunabic Acid, Kaurenic Acid, Kaurenoic Acid, NSC 339145, ent-Kaur-16(17)-en-19-oic Acid, ent-Kaur-16-en-19-oic Acid, (4α)-Kaur-16-en-18-oic Acid
SMILES C=C1C[C@@]23CC[C@]4([H])[C@](C)(C(O)=O)CCC[C@@]4(C)[C@]2([H])CC[C@@H]1C3

Specifications

Quantity 5 mg
InChI Formula InChI=1S/C20H30O2/c1-13-11-20-10-7-15-18(2,16(20)6-5-14(13)12-20)8-4-9-19(15,3)17(21)22/h14-16H,1,4-12H2,2-3H3,(H,21,22)/t14-,15+,16+,18-,19-,20-/m1/s1
Formula Weight 302.2246
Chemical Name or Material ent-Kaurenoic Acid
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.