missing translation for 'onlineSavingsMsg'
Learn More
Learn More
6-Amino-2,4-di-tert-butylphenyl-d9 Methyl Carbonate, TRC
Click to view available options
Quantity:
1 mg
2.5 mg
5 mg
Unit Size:
1mg
2.5mg
5mg
Specifications
Specifications
| Chemical Name or Material | 6-Amino-2,4-di-tert-butylphenyl-d9 Methyl Carbonate |
| Molecular Formula | C32 H31 N2 O3 . Cl O4 |
| Quantity | 2.5 mg |
| Synonym | 2-Amino-4,6-di-tert-butylphenyl-d9 Methyl Carbonate |
| InChI Formula | InChI=1S/C32H30N2O3.ClHO4/c35-32(36)22-10-2-1-9-21(22)27-25-17-19-7-3-13-33-15-5-11-23(28(19)33)30(25)37-31-24-12-6-16-34-14-4-8-20(29(24)34)18-26(27)31;2-1(3,4)5/h1-2,9-10,17-18H,3-8,11-16H2;(H,2,3,4,5) |
| SMILES | OC(=O)c1ccccc1c2c3cc4CCCN5CCCc(c45)c3[o+]c6c7CCCN8CCCc(cc26)c78.[O-]Cl(=O)(=O)=O |
| IUPAC Name | 2-(3-oxonia-9,23-diazaheptacyclo[17.7.1.15,9.02,17.04,15.023,27.013,28]octacosa-1,3,5(28),13,15,17,19(27)-heptaen-16-yl)benzoic acid;perchlorate |
| Molecular Weight (g/mol) | 591.051 |
| Formula Weight | 590.182 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction