missing translation for 'onlineSavingsMsg'
Learn More

6-Chloro-1-benzothiopyran-4-one 1,1-dioxide, 97%, Maybridge™

Product Code. 10161272
Change view
Click to view available options
Quantity:
1 g
10 g
Unit Size:
10g
1g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
10161272 10 g 10g
10425332 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10161272 Supplier Thermo Scientific Maybridge Supplier No. SEW00597EA

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Chemical Identifiers

CAS 90396-06-8
Molecular Formula C9H7ClO3S
Molecular Weight (g/mol) 230.66
MDL Number MFCD00052999
InChI Key ZFZPMYLZPMMUQA-UHFFFAOYSA-N
Synonym 6-chloro-1-benzothiopyran-4-one 1,1-dioxide, 6-chloro-2,3-dihydro-1??-benzothiopyran-1,1,4-trione, maybridge1_007519, 6-chlorothiochroman-4-one 1,1-dioxide, 6-chloro-2h,3h-benzo e thiin-1,1,4-trione, 6-chloro-1,1-dioxo-1lambda6-thiochroman-4-one, 6-chloro-2,3-dihydro-1-benzothiopyran-1,1,4-trione, 6-chloro-2,3-dihydro-4h-thiochromen-4-one 1,1-dioxide, 6-chloro-2,3-dihydro-1, e6-benzothiopyran-1,1,4-trione, 6-chloro-3,4-dihydro-2h-1??-benzothiopyran-1,1,4-trione
PubChem CID 2739106
IUPAC Name 6-chloro-1,1-dioxo-2,3-dihydrothiochromen-4-one
SMILES ClC1=CC=C2C(=C1)C(=O)CCS2(=O)=O

Specifications

CAS Min % 97.0
Assay Percent Range 97%
Quantity 10 g
Formula Weight 230.67
Percent Purity 97%
Chemical Name or Material 6-Chloro-1-benzothiopyran-4-one 1, 1-dioxide
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.