missing translation for 'onlineSavingsMsg'
Learn More

6-Chloro-1-hexanol, 97%

Product Code. 11471988
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
Unit Size:
100g
25g
500g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11471988 500 g 500g
11451988 25 g 25g
11461988 100 g 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11471988 Supplier Thermo Scientific Alfa Aesar Supplier No. A14582.36
Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 2009-83-8 | C6H13ClO | 136.619 g/mol

6-Chloro-1-hexanol is used in the preparation of its corresponding bromide by reaction with sodium bromide using N,N-dimethylformamide and dibromomethane (2:1) as a solvent. It is utilized in the selective elimination of 6-chloro-1-hexene from esters of 6-chloro-1-hexanol.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
6-Chloro-1-hexanol is used in the preparation of its corresponding bromide by reaction with sodium bromide using N,N-dimethylformamide and dibromomethane (2:1) as a solvent. It is utilized in the selective elimination of 6-chloro-1-hexene from esters of 6-chloro-1-hexanol.

Solubility
Soluble in dimethylformamide.

Notes
Incompatible with strong oxidizing agents, strong bases, powdered metal salts, strong reducing agents, acid anhydrides and acid chlorides.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 2009-83-8
Molecular Formula C6H13ClO
Molecular Weight (g/mol) 136.619
MDL Number MFCD00002984
InChI Key JNTPTNNCGDAGEJ-UHFFFAOYSA-N
Synonym 6-chloro-1-hexanol, 6-chlorohexanol, 1-chloro-6-hydroxyhexane, 1-hexanol, 6-chloro, hexamethylene chlorohydrin, 1-chloro-6-hexanol, omega-chlorohexanol, unii-t2hit6n5uf, 6-chloro-hexanol, t2hit6n5uf
PubChem CID 74828
IUPAC Name 6-chlorohexan-1-ol
SMILES C(CCCCl)CCO

Specifications

Density 1.027
Boiling Point 109°C to 111°C (14 mmHg)
Flash Point 98°C (208°F)
Refractive Index 1.456
Quantity 500 g
Beilstein 1697307
Solubility Information Soluble in dimethylformamide.
Formula Weight 136.62
Percent Purity 97%
Chemical Name or Material 6-Chloro-1-hexanol

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.